missing translation for 'onlineSavingsMsg'
Learn More

Dicyclohexano-18-crown-6, mixture of isomers, 97%, Thermo Scientific Chemicals

Product Code. 11374509
Click to view available options
Quantity:
1 g
10 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Dicyclohexano is used as an anion activator and complexing agent which solubilizes alkali metal ions in non-polar solvents. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs etc,.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dicyclohexano is used as an anion activator and complexing agent which solubilizes alkali metal ions in non-polar solvents. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs etc,.

Solubility
Soluble in Toluene. Slightly soluble (13g/L, 26°C) in water.

Notes
Hygroscopic. Store under dry inert gas. Incompatible with strong acids, strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16069-36-6
Molecular Formula C20H36O6
Molecular Weight (g/mol) 372.502
MDL Number MFCD00005099
InChI Key BBGKDYHZQOSNMU-UHFFFAOYSA-N
Synonym dicyclohexano-18-crown-6, dicyclohexyl-18-crown-6, cis-dicyclohexano-18-crown-6, dicyclohexo-18-crown-6, perhydrodibenzo-18-crown-6, dch-18-crown-6, icosahydrodibenzo b,k 1,4,7,10,13,16 hexaoxacyclooctadecine, dicyclohexyl-18-crown-6 ether, dicyclohexyl 18-crown-6, ccris 3612
PubChem CID 85955
SMILES C1CCC2C(C1)OCCOCCOC3CCCCC3OCCOCCO2

Specifications

Melting Point 46°C to 53°C
Flash Point >110°C (230°F)
Odor Odorless
Quantity 10 g
UN Number UN2811
Beilstein 1130529
Sensitivity Hygroscopic
Merck Index 14,2602
Solubility Information Soluble in Toluene. Slightly soluble (13g/L,26°C) in water.
Formula Weight 372.5
Percent Purity 97%
Chemical Name or Material Dicyclohexano-18-crown-6, mixture of isomers
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.