missing translation for 'onlineSavingsMsg'
Learn More
Dicyclohexyltin dibromide, Thermo Scientific™ Product Code.: 11375039

Dicyclohexyltin dibromide, Thermo Scientific™

Product Code. 11375039
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11375039

Brand: Thermo Scientific Alfa Aesar 087800.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2954-94-1
Molecular Formula C12H22Br2Sn
Molecular Weight (g/mol) 444.826
MDL Number MFCD00015370
InChI Key RHFTTZVNBAHQBF-UHFFFAOYSA-L
Synonym dicyclohexyltin dibromide, stannane, dibromodicyclohexyl, dibromo dicyclohexyl stannane, dicyclohexyltindibromide, stannane,dibromodicyclohexylShow MoreShow Less
PubChem CID 97477
IUPAC Name dibromo(dicyclohexyl)stannane
SMILES C1CCC(CC1)[Sn](C2CCCCC2)(Br)Br

Specifications

Melting Point 58°C to 64°C
Color White
Boiling Point ∼150°C (0.3mmHg)
Quantity 5 g
UN Number UN3146
Sensitivity Moisture sensitive
Formula Weight 444.82
Physical Form Crystalline
Chemical Name or Material Dicyclohexyltin dibromide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dicyclohexyltin dibromide, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.