missing translation for 'onlineSavingsMsg'
Learn More
Dicyclopentadiene diepoxide, 98%, Thermo Scientific Chemicals Product Code.: 11357905

Dicyclopentadiene diepoxide, 98%, Thermo Scientific Chemicals

Product Code. 11357905
25 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11357905

Brand: Thermo Scientific Alfa Aesar L02940.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Dicyclopentadiene diepoxide acts as an intermediate for epoxy resins, plasticizers and protective coatings. It finds application as a rubber additive and adhesives, which is used at high temperature. Further, it is used in the preparation of endo-3-hydroxy-exo-9-oxa-exo-tetracyclo[5.3.1.02,6.08,10]undecane using lithium aluminum hydride as a reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dicyclopentadiene diepoxide acts as an intermediate for epoxy resins, plasticizers and protective coatings. It finds application as a rubber additive and adhesives, which is used at high temperature. Further, it is used in the preparation of endo-3-hydroxy-exo-9-oxa-exo-tetracyclo[5.3.1.02,6.08,10]undecane using lithium aluminum hydride as a reagent.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81-21-0
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.204
MDL Number MFCD00077209
InChI Key BQQUFAMSJAKLNB-UHFFFAOYSA-N
Synonym dicyclopentadiene dioxide, dicyclopentadiene diepoxide, unox epoxide 207, epoxide 207, bicyclopentadiene dioxide, unox 207x, unox 207, dicyclopentadiene dioxide van, 1,2:5,6-diepoxyhexahydro-4,7-methanoindan, 4,7-methanoindan, 1,2:5,6-diepoxyhexahydroShow MoreShow Less
PubChem CID 6673
SMILES C1C2C3CC4C(C3C1C5C2O5)O4

Specifications

Melting Point 185°C to 189°C
Density 1.331
Boiling Point 326°C
Odor Odorless
Quantity 25 g
UN Number UN2811
Beilstein 112743
Solubility Information Soluble in water.
Formula Weight 164.2
Percent Purity 99%
Chemical Name or Material Dicyclopentadiene diepoxide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10245292 Certificate TypeCertificate of Analysis Date06/01/2023 Catalog NumberL02940.14,L02940.06
Lot NumberH12U016 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL02940.14,L02940.06
Lot Number10206345 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL02940.14,L02940.06
Lot Number10213877 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL02940.14,L02940.06
Lot Number10220922 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL02940.14,L02940.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dicyclopentadiene diepoxide, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.