Learn More
Hellma™ Didymium Glass F7 Glass Filter
Brand: Hellma™ 666F7-323
This item is not returnable.
View return policy
Description
Order Info
Range
Specifications
DIN EN ISO 17025, US EPA, FDA Regulations | |
Didymium Glass F7 | |
270 to 340 nm |
Glass Filter | |
1 Ea. |