missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethanolamine Buffer Solution pH 10.0, Fisher Chemical™
Click to view available options
Quantity:
1000 mL
Specifications
Specifications
pH | 10.0 |
Physical Form | Liquid |
Quantity | 1000 mL |
Solution Type | Buffer |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
1009389 | Certificate of Analysis | 04/03/2025 | SOL 101 1L |
Safety and Handling


Product Identifier
- Diethanolamine Buffer solution - pH 10.0. European Pharmacopoeia Ref: 4007500. (SOL 101)
- Danger
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity after repeated exposure Category 2
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H373-May cause damage to organs.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P314-Get medical advice/attention if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P501-Dispose of contents/container to .
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Diethanolamine Buffer Solution pH 10.0, Fisher Chemical™ >
Spot an opportunity for improvement?Share a Content Correction