missing translation for 'onlineSavingsMsg'
Learn More

Diethoxydiphenylsilane, 98%, Thermo Scientific Chemicals

Product Code. 11465826
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Reagent for preparation of diphenylsilylene derivatives of diols by exchange.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2553-19-7
Molecular Formula C16H20O2Si
Molecular Weight (g/mol) 272.42
MDL Number MFCD00015126
InChI Key ZZNQQQWFKKTOSD-UHFFFAOYSA-N
Synonym diphenyldiethoxysilane, silane, diethoxydiphenyl, diethoxy diphenyl silane, benzene, 1,1'-diethoxysilylene bis, acmc-1cmgu, diethoxy-di phenyl silane, diethoxydiphenylsilane, zznqqqwfkktosd-uhfffaoysa
PubChem CID 75705
IUPAC Name diethoxy(diphenyl)silane
SMILES CCO[Si](OCC)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Boiling Point 166°C to 168°C (15mmHg)
Quantity 10 g
Beilstein 2281302
Flash Point 175°C (347°F)
Formula Weight 272.42
Percent Purity 98%
Refractive Index 1.5255
Sensitivity Moisture sensitive
Density 1.033
Chemical Name or Material Diethoxydiphenylsilane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.