Learn More
Diethyl 1-benzylpyrrolidine-2,5-dicarboxylate, 97%, Thermo Scientific™
99.80 € - 710.00 €
Chemical Identifiers
CAS | 17740-40-8 |
---|---|
Molecular Formula | C17H23NO4 |
Molecular Weight (g/mol) | 305.374 |
MDL Number | MFCD00830964 |
InChI Key | LDUSEIANLSWKPY-UHFFFAOYSA-N |
Synonym | 2,5-diethyl 1-benzylpyrrolidine-2,5-dicarboxylate, 1-benzyl-pyrrolidine-2,5-dicarboxylic acid diethyl ester, diethyl meso-1-benzyl-2,5-pyrrolidinedicarboxylate, 1-benzyl-2,5-bis ethoxycarbonyl pyrrolidine, diethyl-1-benzylpyrrolidine-2,5-dicarboxylate, pubchem15289, diethyl1-benzylpyrrolidine-2,5-dicarboxylate, hcl, diethyl 1-phenylmethyl pyrrolidine-2,5-dicarboxylate, 1-phenylmethyl pyrrolidine-2,5-dicarboxylic acid diethyl ester |
PubChem CID | 286120 |
IUPAC Name | diethyl 1-benzylpyrrolidine-2,5-dicarboxylate |
SMILES | CCOC(=O)C1CCC(N1CC2=CC=CC=C2)C(=O)OCC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10560922
|
Thermo Scientific Maybridge
KM09423DA |
1 g | Amber Glass Bottle |
99.80 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10242982
|
Thermo Scientific Maybridge
KM09423EA |
10 g | Amber Glass Bottle |
710.00 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemische Identifikatoren
17740-40-8 | |
305.374 | |
LDUSEIANLSWKPY-UHFFFAOYSA-N | |
286120 | |
CCOC(=O)C1CCC(N1CC2=CC=CC=C2)C(=O)OCC |
C17H23NO4 | |
MFCD00830964 | |
2,5-diethyl 1-benzylpyrrolidine-2,5-dicarboxylate, 1-benzyl-pyrrolidine-2,5-dicarboxylic acid diethyl ester, diethyl meso-1-benzyl-2,5-pyrrolidinedicarboxylate, 1-benzyl-2,5-bis ethoxycarbonyl pyrrolidine, diethyl-1-benzylpyrrolidine-2,5-dicarboxylate, pubchem15289, diethyl1-benzylpyrrolidine-2,5-dicarboxylate, hcl, diethyl 1-phenylmethyl pyrrolidine-2,5-dicarboxylate, 1-phenylmethyl pyrrolidine-2,5-dicarboxylic acid diethyl ester | |
diethyl 1-benzylpyrrolidine-2,5-dicarboxylate |
Spezifikation
17740-40-8 | |
95% | |
MFCD00830964 | |
LDUSEIANLSWKPY-UHFFFAOYSA-N | |
diethyl 1-benzylpyrrolidine-2,5-dicarboxylate | |
286120 | |
95% |
95.0 | |
C17H23NO4 | |
2,5-diethyl 1-benzylpyrrolidine-2,5-dicarboxylate, 1-benzyl-pyrrolidine-2,5-dicarboxylic acid diethyl ester, diethyl meso-1-benzyl-2,5-pyrrolidinedicarboxylate, 1-benzyl-2,5-bis ethoxycarbonyl pyrrolidine, diethyl-1-benzylpyrrolidine-2,5-dicarboxylate, pubchem15289, diethyl1-benzylpyrrolidine-2,5-dicarboxylate, hcl, diethyl 1-phenylmethyl pyrrolidine-2,5-dicarboxylate, 1-phenylmethyl pyrrolidine-2,5-dicarboxylic acid diethyl ester | |
CCOC(=O)C1CCC(N1CC2=CC=CC=C2)C(=O)OCC | |
305.374 | |
305.37 | |
Diethyl 1-benzylpyrrolidine-2, 5-dicarboxylate |
Sicherheit und Handhabung
GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.