missing translation for 'onlineSavingsMsg'
Learn More

Diethyl (aminomethyl)phosphonate oxalate, 95%

Product Code. 10663292 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10663292

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 117196-73-3 | C5H14NO3P·C2H2O4 | 257.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 117196-73-3
Molecular Formula C5H14NO3P·C2H2O4
Molecular Weight (g/mol) 257.18
MDL Number MFCD00800255
InChI Key OVHZWLGNOJBTDI-UHFFFAOYSA-N
Synonym diethyl aminomethyl phosphonate oxalate, diethoxyphosphorylmethanamine; oxalic acid, diethyl aminomethyl phosphonate oxalate salt, phosphonic acid, p-aminomethyl-, diethyl ester, ethanedioate 1:1, phosphonic acid, aminomethyl-, diethyl ester, ethanedioate 1:1, acmc-20mn2b, diethyl aminomethylphosphonate oxalate, oxalic acid diethyl aminomethylphosphonate, diethyl aminomethyl phosphonate ethanedioate
PubChem CID 2733594
IUPAC Name diethoxyphosphorylmethanamine;oxalic acid
SMILES CCOP(=O)(CN)OCC.C(=O)(C(=O)O)O

Specifications

Melting Point 118.0°C to 126.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Quantity 1 g
Formula Weight 257.18
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Diethyl (aminomethyl)phosphonate oxalate, 95%

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.