missing translation for 'onlineSavingsMsg'
Learn More

Diethyl dimethylaminomethylenemalonate, 94%, Thermo Scientific Chemicals

Product Code. 11445646
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Derivatives of this compound, diethyl ethoxymethylenemalonate is used in the synthesis of chemotherapeutic agents using 6-aminobenzothiazoles as starting material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Derivatives of this compound, diethyl ethoxymethylenemalonate is used in the synthesis of chemotherapeutic agents using 6-aminobenzothiazoles as starting material.

Solubility
Soluble in methanol.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18856-68-3
Molecular Formula C10H17NO4
Molecular Weight (g/mol) 215.25
MDL Number MFCD00052394
InChI Key XNSXXTKUKYWLAH-UHFFFAOYSA-N
Synonym diethyl dimethylaminomethylenemalonate, diethyl 2-dimethylamino methylidene malonate, diethyl 2-dimethylaminomethylidene propanedioate, diethyl 2-dimethylamino methylene malonate, propanedioic acid, dimethylamino methylene-, diethyl ester, malonic acid, dimethylamino-methyl-, diethyl ester, malonic acid, dimethylaminomethylene-, diethyl ester, 1,3-diethyl 2-dimethylamino methylidene propanedioate, acmc-1buy0
PubChem CID 140439
IUPAC Name diethyl 2-(dimethylaminomethylidene)propanedioate
SMILES CCOC(=O)C(=CN(C)C)C(=O)OCC

Specifications

Boiling Point 134°C to 136°C (0.5 mmHg)
Quantity 5 g
Solubility Information Soluble in methanol.
Formula Weight 215.25
Percent Purity 95%
Chemical Name or Material Diethyl dimethylaminomethylenemalonate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'