missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl ether, anhydrous, ACS, 99% min, stab. with BHT, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
500 g
6 x 500 g
Description
Solvent for U.V. spectrophotometric and it is also important as solvent in production of cellulose plastics such as cellulose acetate. It is also common solvent for Grignard reaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Specifications
Melting Point | 116°C |
Density | 0.713 |
Boiling Point | 34.6°C |
Quantity | 500 g |
Flash Point | -45°C (-49°F) |
Physical Form | Liquid |
Refractive Index | 1.3525 |
Assay Percent Range | 99% min. |
Linear Formula | (C2H5)2O |
UN Number | UN1155 |
Show More |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS02_Extremely_Flammable.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS07_Warning.png)
Product Identifier
- Diethyl ether, anhydrous, ACS
- Danger
- Acute toxicity Category 4
- Flammable liquid Category 1
- Specific target organ toxicity Category 3
- H224-Extremely flammable liquid and vapour.
- H302-Harmful if swallowed.
- H336-May cause drowsiness or dizziness.
- EUH019-May form explosive peroxides.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P240-Ground/bond container and receiving equipment.
- P243-Take precautionary measures against static discharge.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P403+P235-Store in a well-ventilated place. Keep cool.
- MIXTURE LIST-Contains : Ethyl ether
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction