missing translation for 'onlineSavingsMsg'
Learn More
Diethyl Ether, puriss. p.a. ACS Reagent, Honeywell™ Product Code.: 15633680

Diethyl Ether, puriss. p.a. ACS Reagent, Honeywell™

Product Code. 15633680
2.5 L
Click to view available options
Quantity:
500 mL
1 L
2.5 L
5 L
This item is not returnable. View return policy

Product Code. 15633680

Brand: Honeywell Riedel-de Haën 322032.5L

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puriss. p.a., contains BHT as inhibitor, ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥99.8%

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-29-7
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
MDL Number MFCD00011646
InChI Key RTZKZFJDLAIYFH-UHFFFAOYSA-N
Synonym diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
PubChem CID 3283
ChEBI CHEBI:35702
IUPAC Name ethoxyethane
SMILES CCOCC

Specifications

Density 0.706g/mL at 25°C
Quantity 2.5 L
Explosion Limit 0.365
Flash Point −40°C (−40°F)
Ignition Point 320°F
Linear Formula (CH3CH2)2O
UN Number 1155
Beilstein 1696894
Formula Weight 74.12g/mol
Percent Purity ≥99.8%
Grade ACS/puriss. p.a.
Chemical Name or Material Diethyl Ether, Reag. Ph. Eur. reag. ISO
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Diethyl ether
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 1
  • Acute toxicity Category 4
  • Specific target organ toxicity Category 3
Hazard Statement
  • H224-Extremely flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H336-May cause drowsiness or dizziness.
  • EUH019-May form explosive peroxides.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Diethyl Ether, puriss. p.a. ACS Reagent, Honeywell™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.