missing translation for 'onlineSavingsMsg'
Learn More
Diethyl Ether, technical, stabilized, Thermo Scientific Chemicals Product Code.: 13447576
compliance-icons compliance-icons

Diethyl Ether, technical, stabilized, Thermo Scientific Chemicals

Product Code. 13447576
5 L, Metal Drum
Click to view available options
Quantity:
5 L
Packaging:
Metal Drum

Product Code. 13447576

Brand: Thermo Scientific Acros 444260050

Please to purchase this item. Need a web account? Register with us today!

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point -116.0°C
CAS 60-29-7
Density 0.7140g/mL
Boiling Point 34.6°C
Quantity 5 L
Flash Point −45°C
Packaging Metal Drum
Refractive Index 1.3510 to 1.3530
Specific Gravity 0.714
Molecular Formula C4H10O
Linear Formula (CH3CH2)2O
MDL Number MFCD00011646
Viscosity 0.2448 mPa.s (20°C)
Merck Index 15,3861
Solubility Information Solubility in water: 69g/L (20°C). Other solubilities: soluble in concentrated hydrochloric acid,miscible with lower aliphatic alcohols,benzene,,chloroform,petroleum ether and many oils
InChI Key RTZKZFJDLAIYFH-UHFFFAOYSA-N
SMILES CCOCC
IUPAC Name ethoxyethane
Molecular Weight (g/mol) 74.12
Formula Weight 74.12
Grade Technical
Chemical Name or Material Diethyl ether
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0449694 Certificate TypeCertificate of Analysis Date08/03/2024 Catalog Number
Lot NumberA0448821 Certificate TypeCertificate of Analysis Date06/10/2023 Catalog Number
Lot NumberA0398972 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number
Lot NumberA0415980 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number
Lot NumberA0442458 Certificate TypeCertificate of Analysis Date23/05/2022 Catalog Number
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H224-Extremely flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H336-May cause drowsiness or dizziness.
  • EUH019-May form explosive peroxides.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.
  • P243-Take precautionary measures against static discharge.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P403+P235-Store in a well-ventilated place. Keep cool.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Diethyl Ether, technical, stabilized, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.