missing translation for 'onlineSavingsMsg'
Learn More

Diethyl malonate, 99+%, Thermo Scientific Chemicals

Product Code. 10599240
Click to view available options
Quantity:
25 g
500 g
1 kg
2.5 kg
10 kg
Packaging:
Glass bottle
Plastic drum

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105-53-3
Molecular Formula C7H12O4
Molecular Weight (g/mol) 160.17
InChI Key IYXGSMUGOJNHAZ-UHFFFAOYSA-N
Synonym diethyl malonate, malonic ester, propanedioic acid, diethyl ester, dicarbethoxymethane, malonic acid, diethyl ester, carbethoxyacetic ester, malonic acid diethyl ester, 1,3-diethyl propanedioate, ethyl propanedioate, ethyl malonate van
PubChem CID 7761
IUPAC Name diethyl propanedioate
SMILES CCOC(=O)CC(=O)OCC

Specifications

Melting Point -50.0°C
Density 1.0550g/mL
Boiling Point 199.0°C
Flash Point 93°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula C2H5O2CCH2CO2C2H5
Refractive Index 1.4130 to 1.4150
Quantity 500 g
Beilstein 02,573
Fieser 10,182; 13,69
Merck Index 15,3878
Specific Gravity 1.055
Solubility Information Solubility in water: 20g/L (20°C). Other solubilities: soluble in ethanol, ether and acetone
Viscosity 2.14 mPa.s (10°C)
Formula Weight 160.17
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Diethyl malonate
Show More Show Less
Product Identifier
  • Diethyl malonate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'