missing translation for 'onlineSavingsMsg'
Learn More

Diethyl phenylmalonate, 98%, Thermo Scientific Chemicals

Product Code. 10001620
Click to view available options
Quantity:
5 mL
100 mL
250 mL
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83-13-6
MDL Number MFCD00009144
InChI Key FGYDHYCFHBSNPE-UHFFFAOYSA-N
Synonym diethyl phenylmalonate, diethyl 2-phenylmalonate, phenylmalonic acid diethyl ester, propanedioic acid, phenyl-, diethyl ester, diethylphenylmalonate, 1,3-diethyl 2-phenylpropanedioate, malonic acid, phenyl-, diethyl ester, propanedioic acid, 2-phenyl-, 1,3-diethyl ester, diethyl 2-phenylpropane-1,3-dioate, diethyl-phenylmalonate
PubChem CID 66514
IUPAC Name diethyl 2-phenylpropanedioate
SMILES CCOC(=O)C(C1=CC=CC=C1)C(=O)OCC

Specifications

Melting Point 16.0°C to 17.0°C
Color Colorless to Yellow
Density 1.0900g/mL
Boiling Point 170.0°C to 172.0°C (14.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C13H16O4
Linear Formula C2H5O2CCH(C6H5)CO2C2H5
Refractive Index 1.4903 to 1.4923
Quantity 100 mL
Beilstein 09, 854
Specific Gravity 1.09
Solubility Information Solubility in water: immiscible
Molecular Weight (g/mol) 236.27
Formula Weight 236.27
Percent Purity 98%
Physical Form Liquid After Melting
Chemical Name or Material Diethyl phenylmalonate
Show More Show Less
Product Identifier
  • Diethyl phenylmalonate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.