missing translation for 'onlineSavingsMsg'
Learn More
Diethyl phthalate, 99%, Thermo Scientific Chemicals Product Code.: 11418157

Diethyl phthalate, 99%, Thermo Scientific Chemicals

Product Code. 11418157
250 g
Click to view available options
Quantity:
250 g
1000 g
2500 g
This item is not returnable. View return policy

Product Code. 11418157

Brand: Thermo Scientific Alfa Aesar A17529.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Diethyl phthalate is used in the production of perfumes, acetyl salicylic acid, plastics and mosquito repellents. It is used in cosmetic formulations and personal care products such as sprays, nail polishes, and perfumes. It acts as an alcohol denaturant. It is also used as a camphor substitute, plasticizer in solid rocket propellants and wetting agent. It finds application as a surface lubricant in food and pharmaceutical packaging. In addition to this, it is used as a detergent base and aerosol spray.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl phthalate is used in the production of perfumes, acetyl salicylic acid, plastics and mosquito repellents. It is used in cosmetic formulations and personal care products such as sprays, nail polishes, and perfumes. It acts as an alcohol denaturant. It is also used as a camphor substitute, plasticizer in solid rocket propellants and wetting agent. It finds application as a surface lubricant in food and pharmaceutical packaging. In addition to this, it is used as a detergent base and aerosol spray.

Solubility
Miscible with ethanol, chloroform, diethyl ether, benzene, acetone, vegetable oils, ketones, carbon tetrachloride, esters, aromatic hydrocarbons and methanol. Slightly miscible with water and aliphatic solvents.

Notes
Incompatible with strong oxidizing agents, strong acids, permanganates and alkalies. Store in a cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84-66-2
Molecular Formula C12H14O4
Molecular Weight (g/mol) 222.24
MDL Number MFCD00009111
InChI Key FLKPEMZONWLCSK-UHFFFAOYSA-N
Synonym diethyl phthalate, phthalic acid diethyl ester, ethyl phthalate, anozol, neantine, phthalol, solvanol, diethyl o-phthalate, palatinol a, placidol eShow MoreShow Less
PubChem CID 6781
ChEBI CHEBI:34698
IUPAC Name diethyl benzene-1,2-dicarboxylate
SMILES CCOC(=O)C1=CC=CC=C1C(=O)OCC

Specifications

Melting Point -64°C
Density 1.118
Boiling Point 296°C to 298°C
Flash Point 160°C (320°F)
Refractive Index 1.502
Quantity 250 g
Beilstein 1912500
Merck Index 14,7371
Solubility Information Miscible with ethanol,chloroform,diethyl ether,benzene,acetone,vegetable oils,ketones,carbon tetrachloride,esters,aromatic hydrocarbons and methanol. Slightly miscible with water and aliphatic solvents.
Formula Weight 222.24
Percent Purity 99%
Chemical Name or Material Diethyl phthalate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10251514 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA17529.0E,A17529.0B,A17529.30
Lot Number10147659 Certificate TypeCertificate of Analysis Date01/06/2022 Catalog NumberA17529.0E,A17529.0B,A17529.30
Lot Number10141477 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA17529.0E,A17529.0B,A17529.30
Lot Number5008A05N Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA17529.0E,A17529.0B,A17529.30
Lot NumberD3368A Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA17529.0E,A17529.0B,A17529.30
compliance-icons
Product Identifier
  • Diethyl phthalate
Signal Word
  • Warning
Hazard Category
  • Reproductive toxicity Category 2
Hazard Statement
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Diethyl phthalate, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.