missing translation for 'onlineSavingsMsg'
Learn More

Diethyl terephthalate, 95%, Thermo Scientific Chemicals

Product Code. 11449357
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 636-09-9
Molecular Formula C12H14O4
Molecular Weight (g/mol) 222.24
MDL Number MFCD00039891
InChI Key ONIHPYYWNBVMID-UHFFFAOYSA-N
Synonym diethyl terephthalate, p-diethyl phthalate, diethylterephthalate, terephthalic acid, diethyl ester, 1,4-benzenedicarboxylic acid, diethyl ester, diethyl p-phthalate, terephthalic acid diethyl ester, unii-n97x85l3cd, 1,4-diethyl benzene-1,4-dicarboxylate, 1,4-benzenedicarboxylic acid, 1,4-diethyl ester
PubChem CID 12483
IUPAC Name diethyl benzene-1,4-dicarboxylate
SMILES CCOC(=O)C1=CC=C(C=C1)C(=O)OCC

Specifications

Melting Point 44°C to 46°C
Density 1.1
Quantity 50 g
Formula Weight 222.24
Percent Purity 95%
Chemical Name or Material Diethyl terephthalate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'