missing translation for 'onlineSavingsMsg'
Learn More

Diethylaluminium chloride, 0.9M solution in toluene, AcroSeal™, Thermo Scientific Chemicals

Product Code. 10576943
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-10-6, 108-88-3
Molecular Formula C4H10AlCl
Molecular Weight (g/mol) 120.56
MDL Number MFCD00000459
InChI Key YNLAOSYQHBDIKW-UHFFFAOYSA-M
Synonym diethylaluminum chloride, aluminum, chlorodiethyl, diethylaluminium chloride, deac, chlorodiethylaluminum, diethylchloroaluminum, diethylaluminum monochloride, deak, aluminum diethyl monochloride, aluminum, dichlorotetraethyldi
PubChem CID 7277
IUPAC Name chloro(diethyl)alumane
SMILES CC[Al](Cl)CC

Specifications

Color Colorless
Density 0.8760g/mL
Flash Point 4°C
Packaging AcroSeal™ Glass Bottle
Linear Formula ClAl(CH2CH3)2
Quantity 800 mL
Specific Gravity 0.876
Solubility Information Solubility in water: reacts
Formula Weight 120.56
Physical Form Liquid
Chemical Name or Material Diethylaluminium chloride
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Diethylaluminium chloride, 0.9M solution in toluene
Signal Word
  • Danger
Hazard Category
  • Substance which\, in contact with water\, emits flammable gases Category 1
  • Flammable liquid Category 2
  • Aspiration toxicity Category 1
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2
  • Reproductive toxicity Category 2
Hazard Statement
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H225-Highly flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
  • H373-May cause damage to organs.
  • H304-May be fatal if swallowed and enters airways.
  • H336-May cause drowsiness or dizziness.
  • H361d-Suspected of damaging the unborn child.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P402+P404-Store in a dry place. Store in a closed container.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
  • MIXTURE LIST-Contains : Toluene, Diethylaluminium chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.