Learn More
Diethylamine, 99.5%, for analysis, Thermo Scientific Chemicals
27.24 € - 40.50 €
Chemical Identifiers
CAS | 109-89-7 |
---|---|
Molecular Formula | C4H11N |
Molecular Weight (g/mol) | 73.14 |
MDL Number | MFCD00009032 |
InChI Key | HPNMFZURTQLUMO-UHFFFAOYSA-N |
Synonym | diethylamine, n,n-diethylamine, ethanamine, n-ethyl, diethamine, diaethylamin, dwuetyloamina, dietilamina, diethylamin, diethyl amine, n-ethyl-ethanamine |
PubChem CID | 8021 |
ChEBI | CHEBI:85259 |
IUPAC Name | N-ethylethanamine |
SMILES | CCNCC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15562333
|
Thermo Scientific Acros
458490500 |
50 mL |
27.24 €
50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15572333
|
Thermo Scientific Acros
458492500 |
250 mL |
40.50 €
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
109-89-7 | |
73.14 | |
HPNMFZURTQLUMO-UHFFFAOYSA-N | |
8021 | |
N-ethylethanamine |
C4H11N | |
MFCD00009032 | |
diethylamine, n,n-diethylamine, ethanamine, n-ethyl, diethamine, diaethylamin, dwuetyloamina, dietilamina, diethylamin, diethyl amine, n-ethyl-ethanamine | |
CHEBI:85259 | |
CCNCC |
Specifications
109-89-7 | |
0.7100g/mL | |
−23°C | |
C4H11N | |
1.3840 to 1.3860 | |
0.71 | |
Solubility in water: soluble. Other solubilities: miscible with alcohol and most common organic, solvents | |
CCNCC | |
73.14 | |
CHEBI:85259 | |
73.13 | |
Analysis |
-50.0°C | |
55.0°C | |
99.4% min. (GC) | |
(C2H5)2NH | |
MFCD00009032 | |
diethylamine, n,n-diethylamine, ethanamine, n-ethyl, diethamine, diaethylamin, dwuetyloamina, dietilamina, diethylamin, diethyl amine, n-ethyl-ethanamine | |
HPNMFZURTQLUMO-UHFFFAOYSA-N | |
N-ethylethanamine | |
8021 | |
0.34 mPa.s (25°C) | |
99.5% | |
Diethylamine |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.
Highly flammable liquid and vapor.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 203-716-3
- Diethylamine
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 A
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H311-Toxic in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Diethylamine
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.