missing translation for 'onlineSavingsMsg'
Learn More
Diethylaminosulfur trifluoride, 95%, Thermo Scientific Chemicals Product Code.: 10286650

Diethylaminosulfur trifluoride, 95%, Thermo Scientific Chemicals

Product Code. 10286650
25 g, Plastic bottle
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10286650

Brand: Thermo Scientific Acros 216110250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38078-09-0
Molecular Formula C4H10F3NS
Molecular Weight (g/mol) 161.18
MDL Number MFCD00000363
InChI Key CSJLBAMHHLJAAS-UHFFFAOYSA-N
Synonym diethylaminosulfur trifluoride, dast, diethylaminosulphur trifluoride, diethylamino sulfur trifluoride, diethylaminosulfurtrifluoride, diethylaminosulphurtrifluoride, diethylamino sulphur trifluoride, unii-78622bv6ij, sulfur, n-ethylethanaminato trifluoro-, t-4
PubChem CID 123472
IUPAC Name N-ethyl-N-(trifluoro-$l^{4}-sulfanyl)ethanamine
SMILES CCN(CC)S(F)(F)F

Specifications

Color Brown to Yellow
Density 1.3160g/mL
Boiling Point 30.0°C to 32.0°C (3.0 mmHg)
Flash Point 23°C
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Plastic bottle
Linear Formula (C2H5)2NSF3
Refractive Index 1.4100 to 1.4160
Quantity 25 g
Fieser 06,183; 10,142; 12,183; 13,110; 16,128
Merck Index 15, 3124
Specific Gravity 1.316
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in benzene,thf,ether,diglyme and,dichloroethylene
Formula Weight 161.18
Percent Purity 95%
Physical Form Fuming Liquid
Chemical Name or Material Diethylaminosulfur trifluoride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Diethylaminosulfur trifluoride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Self-reactive substance or mixture Type B
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H241-Heating may cause a fire or explosion.
  • H226-Flammable liquid and vapour.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Diethylaminosulfur trifluoride, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.