missing translation for 'onlineSavingsMsg'
Learn More
Diethylmethoxyborane, 1M soln in THF, Thermo Scientific Chemicals Product Code.: 11388136

Diethylmethoxyborane, 1M soln in THF, Thermo Scientific Chemicals

Product Code. 11388136
100 mL
Click to view available options
Quantity:
25 mL
100 mL
This item is not returnable. View return policy

Product Code. 11388136

Brand: Thermo Scientific Alfa Aesar L15107.AE

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7397-46-8
Molecular Formula C5H13BO
Molecular Weight (g/mol) 99.97
MDL Number MFCD00013240
InChI Key FESAXEDIWWXCNG-UHFFFAOYSA-N
Synonym methoxydiethylborane, methyl diethylborinate, diethyl methoxy borane, borinic acid, diethyl-, methyl ester, diethyl methoxyborane, unii-3ah40d7l25, diethyl-methoxyborane, methoxy-diethylborane, methoxydiethyl-borane
PubChem CID 522516
SMILES CCB(CC)OC

Specifications

Density 0.868
Flash Point −6°C (21°F)
Linear Formula (CH3CH2)2BOCH3
Refractive Index 1.404
Quantity 100 mL
UN Number UN2924
Beilstein 2231225
Sensitivity Air and moisture sensitive
IUPAC Name methyl diethylborinate
Formula Weight 99.97
Concentration or Composition (by Analyte or Components) 1M soln in THF
Chemical Name or Material Diethylmethoxyborane
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Diethylmethoxyborane, 1M soln in THF, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.