missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Columbia Agar with 5% Sheep Blood

Artikelnummer. 12800723
missing translation for 'orderingAttributeHoverText'
For Use With (Application):
For the isolation and cultivation of nonfastidious and fastidious microorganisms
missing translation for 'unitSize'
Pack of 20
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 12800723

missing translation for 'mfr': BD 254005

Please to purchase this item. Need a web account? Register with us today!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Recommended when processing clinical specimens for unusual organisms. BD™ BBL™ Columbia Agar with 5% Sheep Blood is a highly nutritious general purpose medium for the isolation and cultivation of nonfastidious and fastidious microorganisms from clinical specimens.

Columbia Agar with 5% sheep blood is a general all-purpose enriched primary isolation medium that allows growth of all clinically significant anaerobes and facultative anaerobes. It is recommended when processing clinical specimens for unusual organisms, and is utilized as the base for media containing blood and for selective media formulations in which various combinations of antimicrobial agents are used as additives.

Columbia Agar with 5% sheep blood derives its superior growth-supporting properties from the combination of two peptones, and yeast extract as a supplier of the B complex vitamins. Corn starch is included to absorb toxic by-products contained in the specimen and serves as an energy source for organisms possessing alphaamylases. Sheep blood allows detection of hemolytic reactions and supplies the X factor (heme) necessary for the growth of many pathogenic species.

  • Colonies tend to be larger and growth is more luxuriant than on media containing other blood agar bases
  • Recommended as a primary isolation medium in the MiQ standards and in other diagnostic manuals
  • In many European countries, this medium has become the most frequently used primary isolation medium for clinical specimens
  • Good cultural response to Streptococcus pyogenes 19615, Streptococcus pneumoniae 6305, Staphylococcus aureus 25923 and Escherichia coli 25922
  • pH (25°C): 7.3 ± 0.2
TRUSTED_SUSTAINABILITY

Spezifikation

Content And Storage Store in the dark at 2–8°C in original wrapping until use.
Format 90mm Stacker™ plates
Media Type Prepared
Product Type Agar
Formulation Pancreatic Digest of Casein 12.0g/L, Peptic Digest of Animal Tissue 5.0g/L, Yeast Extract 3.0g/L, Beef Extract 3.0g/L, Corn Starch 1.0g/L, Sodium Chloride 5.0g/L, Agar 13.5g/L, Defibrinated Sheep Blood 5%
For Use With (Application) For the isolation and cultivation of nonfastidious and fastidious microorganisms
Quantity 20 Each
Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.