missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Dehydrated Culture Media: m Enterococcus Agar

For isolating and enumerating enterococci in water and other materials by membrane filtration or pour plate technique

Brand:  BD 274620


 View more versions of this product

Product Code. 11703493

  • 286.00 € / 500g
Estimated Shipment: 11-10-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Formulation: Tryptose 20g/L, Yeast extract 5g/L, Dextrose 2g/L, Dipotassium Phosphate 4g/L, Sodium Azide 0.4g/L, Agar 10g/L, 2,3,5-Triphenyl Tetrazolium Chloride 0.1g/L

Certifications

CE, ISO, SMWW

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CE, ISO, SMWW
Dehydrated
Agar
For isolating and enumerating enterococci in Water and other materials by membrane filtration or pour plate technique
500 g
<30°C
m Enterococcus Agar
Tryptose 20g/L, Yeast extract 5g/L, Dextrose 2g/L, Dipotassium Phosphate 4g/L, Sodium Azide 0.4g/L, Agar 10g/L, 2,3,5-Triphenyl Tetrazolium Chloride 0.1g/L
Bottle
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3

Hazard Statement
  • H302-Harmful if swallowed.
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P273-Avoid release to the environment.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
  • P501-Dispose of contents/container to in accordance with all local/regional/national/internationalregulations.

Supplemental information
  • MIXTURE LIST-Contains : sodium azide

SDS
Documents

Documents

Certificates
Promotions

Promotions