Learn More
BD Difco™ Disposable Inoculating Needles and Loops
Sterile, disposable inoculating needles do not require flaming
Brand:  BD 220214
 View more versions of this product
Product Code. 10492683
  • 285.00 € / Pack of 250
Estimated Shipment: 16-12-2024
to see stock.
Explore more special offers
Add to basket
Includes: Evans Blue Dye Certificate of Calibration with every box of 1,000 loops, confirming their accuracy and consistency
Description
Description
  • Made from a unique plastic resin for controlled flexibility
  • Resist cutting the agar surface, yet provide sufficient firmness to pick stubborn colonies
  • Free of lubricants, oils and electrostatic charges
  • Color coded
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Single-use
Plastic Resin
Light Green
Evans blue dye certificate of calibration
1 μL
Sterile
Inoculating Loop
250/Pk.