missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Difco™ Soytone

An enzymatic digest of soy.

Brand:  Gibco™ 212488

114.95 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.


 View more versions of this product

Product Code. 16219781

  • 126.00 € / 500g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • An alternative carbohydrate profile to yeast supplements for mammalian cell processes, such as CHO
  • High concentrations of vitamins and carbohydrates
  • Store in cool, dry conditions in well-sealed containers at 2°C to 25°C
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.
Powder
Mammalian Cell Culture
500 g
Mammalian, Insect, Microbial
Yeast Supplement
Difco™
Animal origin-free
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.