Learn More
Masterflex™ Differential Pressure Flowmeters
Brand:  Masterflex™ 32908-69
 View more versions of this product
Product Code. 15305184
  • 1865.00 € / Each
Estimated Shipment: 12-02-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Differential Pressure Flowmeter
145 psig
0.05 to 5 LPM
7 to 30 VDC at 30 mA
50°C
±0.8% (Reading), ±0.2% (Full Scale)
1/8 in. NPT (F)
Versatility and high accuracy from a laminar-based mass flow meter
±0.5%