Learn More
SCHOTT AG Lighting and Imaging™ Diffuse Light Panel for Fiber Optic Flexible Light Guide
Use the Diffuse Light Panel for Fiber Optic Flexible Light Guide for illumination of polished or high reflective surfaces. Additional flexible light guide required (155103 or 155104).
Brand:  SCHOTT AG Lighting and Imaging™ 500100
Product Code. 15783042
  • 500.00 € / 1 unit
Estimated Shipment: 15-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Diffuse and uniform illumination within a certain active area.
  • A white acrylic diffuser plate provides an optimum combination of uniformity and intensity.
  • Mounts directly to respective flexible light guide by means of thumb screw.
  • Housing made of rugged, black anodized aluminum.
  • For use with Cold light source KL1600LED, KL2500LED or KL1500HAL.
  • Dimensions: 137 x 58.5 x 23 mm Housing, 78 x 30 mm Active Area
Specifications
Specifications
Diffuse Facelight
Stereo Microscopy Illumination
2 x M6 holes for articulated arm (158 345)
78 mm
30 mm
137 x 58.5 x 23 mm Housing, 78 x 30 mm Active Area
KL1500 HAL, KL1600 LED, KL2500 LED Cold Light Sources
3.1 in.
1.2 in.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
SCHOTT AG Lighting and Imaging™ Diffuse Light Panel for Fiber Optic Flexible Light Guide > Diffuse Light Panel for Fiber Optic Flexible Light Guide; For Use With: KL1500 HAL, KL1600 LED, KL2500 LED Cold Light Sources