missing translation for 'onlineSavingsMsg'
Learn More

Cytiva DIGE Gel and DIGE Buffer Kit

Tools for increasing the quality, reproducibility, and convenience of 2-D DIGE using Ettan™ DIGE System

437.00 € - 589.00 €

Specifications

Product Type DIGE Gel and Buffer Kit
Form Solid and Liquid
Quantity 1 Kit: 2 x 125 mL anode buffer; 4 x 125 mL cathode buffer; 12 tubes agarose sealing solution
View More Specs

Includes: Kit: 2 x 125mL anode buffer; 4 x 125mL cathode buffer; 12 tubes agarose sealing solution

Products 2
Product Code Brand Color Form Kit Contents Price Quantity & Availability  
Product Code Brand Color Form Kit Contents Price Quantity & Availability  
10310405
View Documents
Cytiva
28-9374-51
Colorless Solid and Liquid Running Buffers and sealing solution for attaching immobiline™ DryStrip gels (ipg strips) to the top of the Polyacrylamide gel
589.00 €
Each
Estimated Shipment: 11-02-2025
to see stock.
Add to basket
 
10636555
View Documents
Cytiva
28-9374-52
Blue Solid and Liquid Running Buffers and sealing solution for attaching immobiline™ DryStrip gels (ipg strips) to the top of the Polyacrylamide Gel, Anode Buffer, cathode Buffer, 12 tubes Agarose sealing solution
437.00 €
1 set
In Stock
to see stock.
Add to basket
 
Description

Description

  • DIGE Gel run with DIGE Buffer Kit results in spot maps with reproducibility compared to homecast polyacrylamide gels, thereby reducing technical variation between gels
  • Ready-to-use
  • Comes with long shelf-life
  • Eliminates the need to make buffers, cast gels and wash glass plates
  • Offers quality and performance of the Laemmli system with reproducibility, convenience and time savings
DIGE Gel
  • Should be used with the DIGE Buffer kit
  • 12.5% precast low fluorscent polyacrylamide gel
  • Cast in low fluorescent glass cassette developed for second dimension 2-D electrophoresis
  • Solid
  • Dimension: 10 x 7.7 x 0.03 in. (255 x 196 x 1mm)
DIGE Buffer Kit
  • Designed two-buffer system for running up to twelve DIGE Gels simultaneously
  • Contains the amount of reagents necessary for a single run of up to 12 DIGE Gels
  • Liquid

Specifications

Specifications

DIGE Gel and Buffer Kit
Solid and Liquid
1 Kit: 2 x 125 mL anode buffer; 4 x 125 mL cathode buffer; 12 tubes agarose sealing solution
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 12 Months

Hazard Statement
  • EUH210-Safety data sheet available on request.

Supplemental information
  • UNKNOWN ACUTE-90 per cent of the mixture consists of component(s) of unknown acute toxicity.
  • UNKNOWN AQUATIC-Contains 90% of components with unknown hazards to the aquatic environment.

SDS
Documents

Documents

Product Certifications