missing translation for 'onlineSavingsMsg'
Learn More

Digi-Sense™ Precalibrated Professional Thermocouple Thermometers

Rugged, heavy-duty construction protects against knocks and jars

Brand:  Digi-Sense™ 20250-18

 View more versions of this product

Product Code. 15800036

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers

Includes: Type K bead wire temperature probe (one for each input), hard carrying case, one 9 V battery, and NIST-traceable calibration report with data supplied by InnoCal. Data logging models 20250-19 and -20 also include USB cable and software.
This item is not returnable. View return policy

Description

Description

Reliable measurements, guaranteed! Precalibrated to NIST-traceable standards|Relative time stamp provides a time reference for major events|Electronic off set function allows compensation of probe errors|Infrared input port allows safe, noncontact measurements with optional IR wand

Services

2 Years

Specifications

Specifications

8 in.
Type K bead wire temperature probe (one for each input), hard carrying case, one 9 V battery, and NIST-traceable calibration report with data supplied by InnoCal.
0.1° 1° above 1000°
None
1372°C
-200°C
-200°C to +1372°C
Backlit LCD
20.32 cm
Type K Thermocouple
2-input Thermocouple Thermometer
2500°F
-328°F
-328°F to +2501°F
Yes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers