missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cole-Parmer™ Digi-Sense™ RTD Extension Cable with 3-Pin Connector
Shop All Cole-Parmer Products
Click to view available options
Length (English):
10 ft.
Unit Size:
Each
Description
- Use adapters or connectors to make your RTD probes compatible with all of our RTD thermometers
- Use the extension cables for remote applications
Specifications
Specifications
| Includes | RTD Extension Cable, 3-Pin Connector Male to Female, 10 ft. FEP Cable |
| Length (Metric) | 3.05 m |
| Material | FEP |
| Length (English) | 10 ft. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction