Learn More
Cole-Parmer™ Digi-Sense™ RTD Wire
Extend the measuring distance of your probes
Brand:  Cole-Parmer™ 08542-31
Product Code. 11795019
  • 188.00 € / Each
Estimated Shipment: 14-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Three-strand RTD wire features excellent resistance to abrasion, chemicals and moisture
  • Each silver-plated copper strad is color-coded with fluorinated ethylene propylene (FEP)
  • Three starnds are twisted to increase flexibility and minimize electromagnetic noise, then jacketed in white FEP
  • FEP insulation provides temperature resistance up to 400°F, and chemical resistance to solvents, acids and oils
  • Additionally, the insulation protects the wire from thermal aging, while maintaining its strength and flexibility
Specifications
Specifications
30.48 m
100 ft.
-200°C to +204°C
PTFE
-328 to +400°F