Learn More
Cole-Parmer™ Digi-Sense™ Detachable Thermocouple Probes
Attach these probes directly to your meter or probe handles. Digi-Sense™ Detachable Thermocouple Probes are ideal for measuring liquids, gases or semisolids.
Brand: Cole-Parmer™ 08117-40
32.75 EUR valid until 2025-03-30
Use promo code "24134" to get your promotional price.
This item is not returnable.
View return policy
Description
Each probe comes with a color coded ANSI mini connector: type J-black, type K-yellow, type T-blue and type E-purple. Response Time: 2 seconds or greater.Specifications
-150°C to +649°C | |
90 mm | |
17 mm |