Learn More
Cole-Parmer™ Digi-Sense™ Traceable™ Triple-display Clock/Timer with Calibration
Use as a digital clock, timer, or stopwatch
Brand:  Cole-Parmer™ 94411-10
Product Code. 13555790
  • 90.70 € / Each
Estimated Shipment: 10-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Get accurate, reliable measurements with included NIST-traceable calibration.
  • Timer displays time of day, or time remaining in any of the three timer chanels.
  • Each channel can be used either simultaneously or independently.
  • Memory recalls previous time set.
  • Display allows simultaneous up and down timing on different channels with unlimited time-outs.
  • Alarm tones are unique for each channel and have adjustable volume.
  • Flashing LCD indicates alarm condition.
  • Traceable™ products are calibrated to NIST-traceable standards and ready to use upon receipt.
  • Each product goes through an intense vetting process before it can carry the Traceable name and it includes NIST-traceable certification.
  • This means that a 17025-accredited calibration test lab has confirmed that the product meets or exceeds all stated specifications, saving you both time and money by not having to calibrate separately.
  • NIST-traceable calibration also helps you meet quality standards and regulatory compliance for ISO, FDA, cGMP, VFC, CAP, CLIA and Joint Commission requirements.
This means that a 17025-accredited calibration test lab has confirmed that the product meets or exceeds all stated specifications, saving you both time and money by not having to calibrate separatelyNIST-traceable calibration also helps you meet quality standards and regulatory compliance for ISO, FDA, cGMP, VFC, CAP, CLIA and Joint Commission requirements
Specifications
Specifications
99 hr. 59 min. 59 sec.
±0.001%
LCD
3
3/8 in.
NIST-traceable
1 min. Continuous Beep and Visible
1 sec.
Traceable™ triple-display clock/timer with calibration
8.3 cm
7.6 cm