Learn More
Cole-Parmer™ CC-200 Stuart™ Digital Colony Counter with IQ/OQ Documents
The CC-200 Stuart™ Digital Colony Counter is a versatile unit can be used with 50 to 90 mm Petri dishes.
Brand:  Cole-Parmer™ 14212-06
Product Code. 15573449
  • 1660.00 € / Each
Estimated Shipment: 23-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Compatible with felt tip markers for pressure-sensitive counting
  • Read individual plates or calculate an average over multiple plates
  • Energy-saving bright-white, LED light source
  • Antimicrobial coating provides additional protection
  • Audible count confirmation, adjustable pressure sensitivity, and option for light/dark background increases flexibility
  • Connects to a printer or computer to record individual or average plate counts
  • Includes IQ/OQ documents
  • 3 year warranty
  • Previous part number SC6PLUS/IQOQ
This colony counter registers a count each time you press a felt tip marker to the Petri dish. Cumulative count may be read on the digital display and is confirmed by an audible tone (can be turned on or off). Adjust the pressure required to measure a count — suitable to each user.
Sub-stage illumination is provided by energy-saving, bright LEDs for glare-free observation. Enhance counting of translucent or difficult-to-see colonies with the supplied black background filter. Calculate the average colony count over multiple plates using the averaging facility. Count results and statistics, including standard deviation, can be sent directly to a printer or computer using the supplied USB cable. Order the 1.7x and 3x magnification adapters separately for easier viewing of small colonies.
Please Note: Effective June 2022, the Stuart Digital Colony Counter Model number SC6PLUS/IQOQ has now been rebranded Cole-Parmer under Model number CC-200. All the same great features and specifications, just with a new and improved look.
Specifications
Specifications
Colony Counter (Digital)
5.5 in.
11.8 in.
12.2 in.
50/60 Hz
Black Background Filter, Wolffhuegel Graticules (2), 50- to 90-mm Dish Centering Adapters (2), and UK/Shuko Power Plug Adapters
30 x 31 x 14 cm
14 cm
30 cm
31 cm
90/230 VAC
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products