missing translation for 'onlineSavingsMsg'
Learn More

Gibertini Elettronica™ Digital Contact Angle

Measure liquid surface tension with efficiency, using this digital tensiometer. The Gibertini Elettronica™ Digital Contact Angle features a simple, robust design for fast response times and accurate results.

Brand:  Gibertini Elettronica™ DCA400

Encompass_Preferred

Product Code. 11920224

  • 21039.48 € / Each
Estimated Shipment: 03-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Display of surface tension value expressed in mN/m (dyne/cm) measured with the Wilhelmy method
  • Autocalibration with internal mass
  • Contact angle 0 to 180 degrees
  • Glass plates (dimensions 24 x 24 x 0.15mm)
  • Suspended system for the arrangement of glass plate
  • Glass container for liquid sample
  • Manual adjustable lab jack
  • Automatic jack with programmable speed (from 21 to 328 mm/sec)
  • Platinum Wilhelmy plate
  • Platinum Du Nouy ring
  • Floater calibrated in weight and in volume and double wall cylinder for relative density measurement:
  • Specific container for sample thermostatation
  • Special pan for metrological control
  • Thermometric probe PT 100 1/3 DIN
Includes software with the following functionality:
  • Contemporaneous display on PC
  • Force changes during the measurements
  • Past time
  • Surface tension value
  • Data recording (useful for other elaborations)
  • Changing value of perimeter plate (if different from the standard)
  • Selection of manual or automatic system for the end of measurement determination
Specifications

Specifications

Digital Tensiometer
Measuring liquid surface tension
Glass plate dimensions: 24 x 24 x 0.15mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
Gibertini Elettronica™ Digital Contact Angle

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.