missing translation for 'onlineSavingsMsg'
Learn More

Grant Instruments™ T100 Series Digital Heated Circulating Bath, Stainless Steel Tank

Economy model with digital thermostatic control unit and stainless tank for straightforward applications requiring accurate temperature control.

Brand:  Grant Instruments™ T100-ST26

1752.67 EUR valid until 2024-12-31
Use promo code "21916" to get your promotional price.


 View more versions of this product

Product Code. 12787809

This item is not returnable. View return policy

Description

Description

  • Stability ±0.05ºC.
  • Clear, bright 4 digit LED display
  • Simple, intuitive user interface: dial and two function buttons.
  • Model available with/without clamp (T-Clamp)
  • Low-liquid detection (float switch)
  • Fixed over temperature
  • 2 point user calibration
  • Visual alarm
Specifications

Specifications

26 L
No
540 x 330 x 405 mm
0°C to 100°C
Stainless Steel Tank
120/230 V
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers