Learn More
SI Analytics™ Digital Multi-Parameter Portable Meter
SI Analytics™ Digital Multi-Parameter Portable Meter has a single cast silicone keypad that can be cleaned easily with a soft brush under a water jet.
Brand: SI Analytics™ 2FD354
1819.25 EUR valid until 2024-12-31
Use promo code "21658" to get your promotional price.
This item is not returnable.
View return policy
Description
- Parameters: pH, mV, O2 (saturation, concentration, partial pressure), Conductivity (spec. resistance, salinity, TDS), Temperature, Turbidity
- 1 universal measuring channel
- Protection class IP 67
- Two field-suitable waterproof USB interfaces for connecting memory sticks or selected printers and for data transfer to PC
Specifications
Manual, time controlled data logger | |
Graphic, BW | |
Multi 3510 IDS SET4 |
Mini USB-B interface | |
Manual: 500 data sets; Automatic: 4500 data sets | |
4 alkaline batteries, USB |