Learn More
TFA Dostmann Digital Stopwatch
This small and compact digital stopwatch is the ideal companion for leisure and work. It reliably measures the time to 1/100 second accuracy and can show split times. The stopwatch has useful additional functions, such as an hourly chime or an alarm with snooze function.
Brand:  TFA Dostmann 38.2030
Product Code. 17971652
  • 10.00 € / Each
Explore more special offers
Description
Description
This small and compact stopwatch is the ideal companion for leisure and work. Whether in competition, training or in the lab, it reliably measures the time to 1/100 second accuracy and can show split times. What's more, it has useful additional functions, such as an hourly chime or an alarm with snooze function. A practical neck strap is included.
  • 1/100 seconds
  • Split time
  • Clock, date with weekday, hourly chime, alarm with snooze function
  • Lanyard
Specifications
Specifications
Stopwatch
Audible/Visible
50 g
23 hr. 59 min. 59 sec.
LCD