Learn More
Fisherbrand™ Digital Thermometers with Stainless-Steel Probe on Cable
Easy-to-read digital thermometers are compact to hold in your hand or mount on wall in restricted areas
Brand:  Fisherbrand™ 93677-09
 View more versions of this product
Product Code. 11779725
  • 102.00 € / Each
Explore more special offers
Description
Description
Easy-to-read digital thermometers are compact to hold in your hand or mount on wall in restricted areas. Fisher Scientific™ Digital thermometer with stainless-steel probe is chemically resistant to most acids, bases and solvents and may be used to measure temperature of gases, liquids, food, and other semisolids.
  • Models 15-077-16, -15, -21, -8B, -29 and 14-648-46 have alarm that can be set to alert user to temperatures that exceed setpoints
  • Some models (such as 15-077-16 and -15) have special features that print temperature, high/low memory and time of day; 15-077-29 thermometer has additional timer alarms
  • For a miniature-size thermometer, see model 15-077-32  
For use in cuvets, soil, incubators, petroleum products and piercing food samples. Waterproof thermometers (models 15-077-9E, -9D and 14-648-46) are ideal for wet labs, wash-down areas and outdoor applications in the agricultural, petrochemical, food, and pharmaceutical industries.
Certifications
Individually serial-numbered Traceable Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™; indicates traceability to standards provided by NIST™.
Specifications
Specifications
ISO 17025, A2LA, NIST
4 x 3.25 x 4.75 in. (10 x 8.2 x 12.06 cm)
4.75 in.
Traceable certificate, Stainless Steel probe, cable and Silver Oxide battery
7.25 in.
73.7 cm
0.1°C
110 g
Traceable Digital Thermometer w/Probe
302°F
-58°F
-58°F to +302°F
±1°C from -20° to +100°C
8.2 cm
0.19 in.
Wet labs, wash down areas and outdoor applications in the agricultural, petrochemical, food and pharmaceutical industries
12.06 cm
29 in.
100 mm
18.41 cm
4 oz.
3.25 in.
4 in.
150°C
-50°C
-50°C to +150°C
Digital LCD