missing translation for 'onlineSavingsMsg'
Learn More

Digoxin, 95%, Thermo Scientific Chemicals

Product Code. 10460501
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20830-75-5
Molecular Formula C41H64O14
Molecular Weight (g/mol) 780.94
MDL Number MFCD00003674
InChI Key LTMHDMANZUZIPE-PUGKRICDSA-N
Synonym digoxin, 12beta-hydroxydigitoxin, digoxine, lanoxin, lanoxicaps, digosin, digossina, digoxina, digoxinum, lanoxicaps tn
PubChem CID 2724385
ChEBI CHEBI:4551
IUPAC Name 3-[(3S,5R,8R,9S,10S,12R,13S,14S,17R)-3-[(2R,4S,5S,6R)-5-[(2S,4S,5S,6R)-5-[(2S,4S,5S,6R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tet
SMILES CC1C(C(CC(O1)OC2C(OC(CC2O)OC3C(OC(CC3O)OC4CCC5(C(C4)CCC6C5CC(C7(C6(CCC7C8=CC(=O)OC8)O)C)O)C)C)C)O)O

Specifications

Melting Point 248.0°C to 250.0°C
Assay Percent Range 94% min. (on dried substance) (HPLC)
Packaging Glass bottle
Quantity 1 g
Merck Index 15,3186
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in diluted alcohol,pyridine,mixture of,chloroform and alcohol,almost insoluble in ether,acetone,ethylacetate,,chloroform
Formula Weight 780.94
Percent Purity 95%
Chemical Name or Material Digoxin

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'