missing translation for 'onlineSavingsMsg'
Learn More

Digoxin, 96%, Thermo Scientific Chemicals

Product Code. 11476400
Click to view available options
Quantity:
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

 

Digoxin is used in several cardiac pathologies such as atrial fibrillation and heart failure.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Digoxin is used in several cardiac pathologies such as atrial fibrillation and heart failure.

Solubility
Soluble in dimethyl sulfoxide and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Specifications

CAS 20830-75-5
Melting Point ∼250°C (decomposition)
Molecular Formula C41H64O14
MDL Number MFCD00003674
Quantity 25 g
UN Number UN2811
Merck Index 14,3167
Synonym digoxin, 12 beta-hydroxydigitoxin, digoxine, lanoxin, lanoxicaps
Solubility Information Soluble in dimethyl sulfoxide and methanol.
InChI Key LTMHDMANZUZIPE-PUGKRICDSA-N
SMILES C[C@H]1O[C@H](C[C@H](O)[C@@H]1O)O[C@H]1[C@@H](O)C[C@H](O[C@H]2[C@@H](O)C[C@H](O[C@H]3CC[C@@]4(C)[C@H](CC[C@@H]5[C@@H]4C[C@@H](O)[C@]4(C)[C@H](CC[C@]54O)C4=CC(=O)OC4)C3)O[C@@H]2C)O[C@@H]1C
IUPAC Name 4-[(1R,3aS,3bR,5aR,7S,9aS,9bS,11R,11aS)-7-{[(2R,4S,5S,6R)-5-{[(2S,4S,5S,6R)-5-{[(2S,4S,5S,6R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy}-4-hydroxy-6-methyloxan-2-yl]oxy}-4-hydroxy-6-methyloxan-2-yl]oxy}-3a,11-dihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]-2,5-dihydrofuran-2-one
Molecular Weight (g/mol) 780.95
PubChem CID 2724385
ChEBI CHEBI:4551
Formula Weight 780.95
Percent Purity 96%
Chemical Name or Material Digoxin
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.