missing translation for 'onlineSavingsMsg'
Learn More
Dihydroergocristine methanesulfonate, Thermo Scientific Chemicals Product Code.: 15443379

Dihydroergocristine methanesulfonate, Thermo Scientific Chemicals

Product Code. 15443379
1 g
Click to view available options
Quantity:
200 mg
1 g
This item is not returnable. View return policy

Product Code. 15443379

Brand: Thermo Scientific Alfa Aesar J60638.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

5-HT receptor antagonist; partial agonist at adrenergic and dopaminergic receptors

5-HT receptor antagonist; partial agonist at adrenergic and dopaminergic receptors

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24730-10-7
Molecular Formula C36H45N5O8S
Molecular Weight (g/mol) 707.843
MDL Number MFCD00153792
InChI Key SPXACGZWWVIDGR-SPZWACKZSA-N
Synonym dihydroergocristine mesylate, dihydroergocristinmesilat, unii-ds7cl18uam, dihydroergocristine mesilate, 9,10-dihydroergocristine methanesulfonate, ds7cl18uam, dihydroergocristine methanesulfonate, dihydroergocristine methanesulfonate salt, 5'alpha-benzyl-9,10alpha-dihydro-12'-hydroxy-2'-isopropylergotaman-3',6',18-trione monomethanesulphonate, diertine tn
PubChem CID 444034
ChEBI CHEBI:31490
SMILES CC(C)C1(C(=O)N2C(C(=O)N3CCCC3C2(O1)O)CC4=CC=CC=C4)NC(=O)C5CC6C(CC7=CNC8=CC=CC6=C78)N(C5)C.CS(=O)(=O)O

Specifications

Color White
Quantity 1 g
Merck Index 14,3653
Formula Weight 707.8
Physical Form Powder
Chemical Name or Material Dihydroergocristine methanesulfonate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN03J001 Certificate TypeCertificate of Analysis Date09/02/2023 Catalog NumberJ60638.!3,J60638.03
Lot NumberM25J024 Certificate TypeCertificate of Analysis Date30/01/2023 Catalog NumberJ60638.!3,J60638.03
Lot NumberZ29H024 Certificate TypeCertificate of Analysis Date17/11/2022 Catalog NumberJ60638.!3,J60638.03
Lot NumberQ27I033 Certificate TypeCertificate of Analysis Date05/05/2022 Catalog NumberJ60638.!3,J60638.03
Lot NumberW26F014 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberJ60638.!3,J60638.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dihydroergocristine methanesulfonate, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.