missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dihydrogen dinitrosulfatoplatinate(II) solution, Pt 4-6% (cont. Pt), Thermo Scientific Chemicals
Click to view available options
Quantity:
∼0.5 g
∼10 g
∼2 g
Description
Dihydrogen dinitrosulfatoplatinate(II) solution is used in the preparation of electrodeposition of platinum films and biocompatible platinum thin film electrodeposition on tin substrates, which acts as the interconnects material for implantable medical applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Color | Brown |
Physical Form | Liquid |
Quantity | ∼0.5 g |
Solubility Information | Miscible with water. |
Formula Weight | 384.17 (Anhydrous) |
Appearance | Brown liquid |
Chemical Name or Material | Dihydrogen dinitrosulfatoplatinate(II) solution, Pt 4-6% (cont. Pt) |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'