Learn More
Dihydrogen hexachloroiridate(IV) hydrate, 99% (metals basis), Ir 38-42%, Thermo Scientific Chemicals
314.00 € - 5804.00 €
Chemical Identifiers
CAS | 110802-84-1 |
---|---|
Molecular Formula | Cl6H2Ir |
Molecular Weight (g/mol) | 406.93 |
MDL Number | MFCD00149906 |
InChI Key | IBMVGQBDTOACBV-UHFFFAOYSA-J |
Synonym | Hydrogen hexachloroiridate(IV); Chloroiridic acid |
PubChem CID | 16211483 |
SMILES | [H+].[H+].Cl[Ir+4](Cl)(Cl)(Cl)(Cl)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11311658
|
Thermo Scientific Alfa Aesar
011031.03 |
1 g |
314.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11321658
|
Thermo Scientific Alfa Aesar
011031.06 |
5 g |
1479.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11301658
|
Thermo Scientific Alfa Aesar
011031.14 |
25 g |
5804.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Dihydrogen hexachloroiridate(IV) hydrate is involved in the electrochemical synthesis of polyaniline on the surface of non-noble metal electrodes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
110802-84-1 | |
406.93 | |
IBMVGQBDTOACBV-UHFFFAOYSA-J | |
16211483 |
Cl6H2Ir | |
MFCD00149906 | |
Hydrogen hexachloroiridate(IV); Chloroiridic acid | |
[H+].[H+].Cl[Ir+4](Cl)(Cl)(Cl)(Cl)Cl |
Specifications
110802-84-1 | |
Lump | |
MFCD00149906 | |
Hydrogen hexachloroiridate(IV); Chloroiridic acid | |
IBMVGQBDTOACBV-UHFFFAOYSA-J | |
tetrachloroiridium;hydrate;dihydrochloride | |
16211483 | |
99% | |
(metals basis) |
Black | |
Cl6H2Ir | |
UN3260 | |
Soluble in alcohol and water | |
[H+].[H+].Cl[Ir+4](Cl)(Cl)(Cl)(Cl)Cl | |
406.93 | |
406.93 (Anhydrous) | |
Ir 38 to 42% | |
Dihydrogen hexachloroiridate(IV) hydrate |
Safety and Handling
GHS H Statement
H314-H318-H290-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
May be corrosive to metals.
Harmful if swallowed.
P234-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P390-P501c
H290-H302-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
missing translation for 'einecsNumber' : 241-012-8
missing translation for 'rtecsNumber' : NO3592500
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only