missing translation for 'onlineSavingsMsg'
Learn More

Dihydrogen hexachloroplatinate(IV) hydrate, 99.9% (metals basis), Thermo Scientific Chemicals

For Catalysis, Electroplating, pHotography

115.58 € - 1568.81 €

Chemical Identifiers

CAS 26023-84-7
Molecular Formula Cl6H2Pt
Molecular Weight (g/mol) 409.80
MDL Number MFCD00149909
InChI Key ZKOQTCCVSRSECD-UHFFFAOYSA-J
Synonym CPA, Chloroplatinic acid
IUPAC Name dihydrogen hexachloroplatinumtetrakis(ylium)
SMILES [H+].[H+].Cl[Pt+4](Cl)(Cl)(Cl)(Cl)Cl
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11351698
View Documents Promotion Details
Thermo Scientific Alfa Aesar
011051.03
1 g
115.58 €
1g
Please to purchase this item. Need a web account? Register with us today!
11361698
View Documents Promotion Details
Thermo Scientific Alfa Aesar
011051.06
5 g
428.13 €
5g
Please to purchase this item. Need a web account? Register with us today!
11371698
View Documents Promotion Details
Thermo Scientific Alfa Aesar
011051.14
25 g
1568.81 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Catalysis, electroplating, photography, platinum mirrors, printing for etching of zinc, producing fine color in glass and porcelain, indelible ink, microscopy.Dihydrogen hexachloroplatinate(IV) hexahydrate is used for the determination of potassium. It is also used for the purification of platinum by treating it with ammonium chloride followed by heating of ammonium hexachloroplatinate with hydrogen yields elemental platinum. It also finds application in catalysis, electroplating, photography, platinum mirrors, printing for etching of zinc, producing fine color in glass and porcelain, indelible ink and microscopy. It is a precursor for the preparation of other platinum catalysts. In addition to this, it is also used as an analytical reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

26023-84-7
409.80
ZKOQTCCVSRSECD-UHFFFAOYSA-J
dihydrogen hexachloroplatinumtetrakis(ylium)
Cl6H2Pt
MFCD00149909
CPA, Chloroplatinic acid
[H+].[H+].Cl[Pt+4](Cl)(Cl)(Cl)(Cl)Cl

Specifications

60°C (decomposition)
Orange to White
99.9% (metals basis)
MFCD00149909
Soluble in water,ethanol,ether,ethyl acetate and acetone. Insoluble in nitric acid.
[H+].[H+].Cl[Pt+4](Cl)(Cl)(Cl)(Cl)Cl
409.80
Hygroscopic
Dihydrogen hexachloroplatinate (IV) hexahydrate
26023-84-7
Crystalline
Cl6H2Pt
CPA, Chloroplatinic acid
ZKOQTCCVSRSECD-UHFFFAOYSA-J
dihydrogen hexachloroplatinumtetrakis(ylium)
409.82 (Anhydrous)
2.43
Videos
Safety and Handling

Safety and Handling

P260-P264b-P270-P271-P272-P280-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c

H301-H314-H317-H334-H335

missing translation for 'dotInformation' : DOT Class: 8, Packaging Group: III

missing translation for 'einecsNumber' : 607-848-6

missing translation for 'rtecsNumber' : TP1500000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Certificates

RUO – Research Use Only