missing translation for 'onlineSavingsMsg'
Learn More

Dihydrojasmone, 97%, Thermo Scientific Chemicals

Product Code. 11449556
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1128-08-1
Molecular Formula C11H18O
Molecular Weight (g/mol) 166.26
MDL Number MFCD00036480
InChI Key YCIXWYOBMVNGTB-UHFFFAOYSA-N
Synonym dihydrojasmone, 3-methyl-2-pentylcyclopent-2-enone, jasmone, dihydro, 2-cyclopenten-1-one, 3-methyl-2-pentyl, 2-pentyl-3-methyl-2-cyclopenten-1-one, dihydrojasmone natural, 3-methyl-2-n-pentanyl-2-cyclopenten-1-one, 2-amyl-3-methyl-2-cyclopenten-1-one, unii-y953r7pp90, fema no. 3763
PubChem CID 62378
IUPAC Name 3-methyl-2-pentylcyclopent-2-en-1-one
SMILES CCCCCC1=C(C)CCC1=O

Specifications

Density 0.91
Boiling Point 120°C to 121°C (12 mmHg)
Flash Point 121°C (250°F)
Refractive Index 1.479
Quantity 100 g
Beilstein 1906471
Formula Weight 166.26
Percent Purity 97%
Chemical Name or Material Dihydrojasmone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.