missing translation for 'onlineSavingsMsg'
Learn More

Dihydrokainic acid, Tocris Bioscience™

Product Code. 18054342
Click to view available options
Quantity:
1 mg
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

EAAT2 (GLT-1)-selective non-transportable inhibitor of L-glutamate and L-aspartate uptake

Dihydrokainic acid is an EAAT2(GLT1)-selective non-transportable inhibitor of L-glutamate and L-aspartate uptake (Ki = 23 μM). 130-fold selective over EAAT1 and EAAT3 (Ki > 3 mM).

Chemical Identifiers

CAS 52497-36-6
Molecular Formula C10H17NO4
Molecular Weight (g/mol) 215.25
MDL Number MFCD03412037
InChI Key JQPDCKOQOOQUSC-OOZYFLPDSA-N
Synonym dihydrokainic acid, dihydrokainate, dihydrokainic acid, natural, 2s,3s,4r-2-carboxy-4-isopropyl-3-pyrrolidineacetic acid, 3s,4r-3-carboxymethyl-4-propan-2-yl-l-proline, 2s,3s,4r-2-carboxy-4-1-methylethyl-3-pyrrolidineacetic acid, 2s,3s,4r-3-carboxymethyl-4-propan-2-yl pyrrolidine-2-carboxylic acid, 2s,3s,4r-3-carboxymethyl-4-isopropylpyrrolidine-2-carboxylic acid, 1ftk, 1xhy
PubChem CID 107883
ChEBI CHEBI:43562
IUPAC Name (2S,3S,4R)-3-(carboxymethyl)-4-(propan-2-yl)pyrrolidine-2-carboxylic acid
SMILES CC(C)[C@H]1CN[C@@H]([C@H]1CC(O)=O)C(O)=O

Specifications

Quantity 1 mg
Formula Weight 215.25
Percent Purity >98%
Chemical Name or Material Dihydrokainic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.