missing translation for 'onlineSavingsMsg'
Learn More
Diiodomethane, 99+%, stabilized with silver wire, Thermo Scientific Chemicals Product Code.: 10160312

Diiodomethane, 99+%, stabilized with silver wire, Thermo Scientific Chemicals

Product Code. 10160312
25 g, Glass bottle
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10160312

Brand: Thermo Scientific Acros 169830250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-11-6, 7440-22-4
Molecular Formula CH2I2
Molecular Weight (g/mol) 267.84
MDL Number MFCD00001079
InChI Key NZZFYRREKKOMAT-UHFFFAOYSA-N
Synonym methylene iodide, methane, diiodo, methylene diiodide, mi-gee, dijodmethan, methylenjodid, dijodmethan czech, methylenjodid czech, di-iodomethane, diiodo-methane
PubChem CID 6346
IUPAC Name diiodomethane
SMILES ICI

Specifications

Melting Point 6.0°C
Color Red to Yellow
Density 3.3200g/mL
Boiling Point 181.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula CH2I2
Refractive Index 1.737
Quantity 25 g
Beilstein 01,71
Fieser 09,170; 11,182; 17,116
Merck Index 15,6137
Specific Gravity 3.32
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with alcohol,propanol,isopropanol,,hexane,cyclohexane,ether,chloroform and benzene
Stabilizer Silver wire
Viscosity 2.80 mPa.s (20°C)
Formula Weight 267.83
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Diiodomethane
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0458435 Certificate TypeCertificate of Analysis Date16/12/2024 Catalog Number169830025,169835000,169830250,169831000
Lot NumberA0457183 Certificate TypeCertificate of Analysis Date28/10/2024 Catalog Number169830025,169835000,169830250,169831000
Lot NumberA0463549 Certificate TypeCertificate of Analysis Date19/06/2024 Catalog Number169830025,169835000,169830250,169831000
Lot NumberA0461217 Certificate TypeCertificate of Analysis Date18/03/2024 Catalog Number169830025,169835000,169830250,169831000
Lot NumberA0458435 Certificate TypeCertificate of Analysis Date18/12/2023 Catalog Number169830025,169835000,169830250,169831000
compliance-icons compliance-icons
Product Identifier
  • Diiodomethane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Dijodmethan

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Diiodomethane, 99+%, stabilized with silver wire, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.