missing translation for 'onlineSavingsMsg'
Learn More

Diisobutylaluminium hydride, 1.1M solution in cyclohexane, AcroSeal™, Thermo Scientific Chemicals

Brand:  Thermo Scientific Acros 201038000

276.45 EUR valid until 2024-12-31
Use promo code "21665" to get your promotional price.



 View more versions of this product

Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Product Code. 10747213

Quantity Price
1 291.00 € / 800mL
6 230.76 € / 800mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers


This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 
This item is not returnable. View return policy

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid, Reduction reagent
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Diisobutylaluminium hydride
1.1M Solution in Cyclohexane
-70.0°C
[(CH3)2CHCH2]2AIH
04, IV, 4400
01,260; 02,140; 03,101; 04,158; 05,224; 06,198; 07,111; 08,173; 09,171; 10,149; 11,185; 12,191; 13,115; 14,138; 15,137; 16,27; 17,123
15, 3212
Solubility in water: vigorous reaction. Other solubilities: miscible with saturated aliphatic and aromatic, hydrocarbons
AcroSeal™ Glass Bottle
800 mL
Solution
Undesignated
1191-15-7, 110-82-7
C8H19Al
MFCD00008928
19.5 to 20.5% (DIBAL-H, on Al basis)
0.7800g/mL
DIBAL-H, 1.1M solution in cyclohexane
142.22
0.78
−018°C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Diisobutylaluminium hydride, 1.1M solution in cyclohexane

Signal Word
  • Danger

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity Category 3
  • Substance which\, in contact with water\, emits flammable gases Category 1

Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H314-Causes severe skin burns and eye damage.
  • H336-May cause drowsiness or dizziness.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P331-Do NOT induce vomiting.
  • P402+P404-Store in a dry place. Store in a closed container.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only