Learn More
Diisobutylaluminium hydride, 1.2M (20 wt%) solution in toluene, AcroSeal™, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 201084000
133.78 EUR valid until 2024-12-31
Use promo code "21665" to get your promotional price.
View more versions of this product
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid, Reduction reagentSpecifications
Diisobutylaluminium hydride | |
0.8480g/mL | |
4°C | |
C8H19Al | |
MFCD00008928 | |
04, IV, 4400 | |
15, 3212 | |
DIBAL-H | |
142.22 | |
Solution |
1191-15-7, 108-88-3 | |
110.0°C | |
AcroSeal™ Glass Bottle | |
[(CH3)2CHCH2]2AIH | |
400 mL | |
01,260; 02,140; 03,101; 04,158; 05,224; 06,198; 07,111; 08,173; 09,171; 10,149; 11,185; 12,191; 13,115; 14,138; 15,137; 16,27; 17,123 | |
0.848 | |
Solubility in water: reacts violently. Other solubilities: miscible with saturated aliphatic and aromatic,hydrocarbons | |
142.22 |
Safety and Handling
- Diisobutylaluminium hydride, 20 wt% solution in toluene
Signal Word
- Danger
Hazard Category
- Aspiration toxicity Category 1
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 1 A
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
- Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
- H304-May be fatal if swallowed and enters airways.
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H361d-Suspected of damaging the unborn child.
- H373-May cause damage to organs.
- H336-May cause drowsiness or dizziness.
- H260-In contact with water releases flammable gases which may ignite spontaneously.
- H225-Highly flammable liquid and vapour.
- EUH014-Reacts violently with water.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P243-Take precautionary measures against static discharge.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P331-Do NOT induce vomiting.
- P402+P404-Store in a dry place. Store in a closed container.
- P403+P235-Store in a well-ventilated place. Keep cool.
RUO – Research Use Only