missing translation for 'onlineSavingsMsg'
Learn More

Diisobutylamine, 99%, Thermo Scientific Chemicals

Product Code. 10583131
Click to view available options
Quantity:
500 mL
1 L
2.5 L
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-96-3
Molecular Formula C8H20N
Molecular Weight (g/mol) 130.25
MDL Number MFCD00008930
InChI Key OBYVIBDTOCAXSN-OCAPTIKFSA-O
Synonym diisobutylamine, 1-propanamine, 2-methyl-n-2-methylpropyl, bis 2-methylpropyl amine, amine, diisobutyl, bis beta-methylpropyl amine, di-isobutylamine, n,n-bis 2-methylpropyl amine, unii-t18y0a819s, ccris 6232, di-2-methylpropyl amine
PubChem CID 8085
IUPAC Name 2-methyl-N-(2-methylpropyl)propan-1-amine
SMILES CC[C@H](C)[NH2+][C@H](C)CC

Specifications

Melting Point -77°C
Density 0.7400g/mL
Boiling Point 137°C to 139°C
Flash Point 26°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula [(CH3)2CHCH2]2NH
Refractive Index 1.4071 to 1.4091
Quantity 500 mL
Beilstein 04, 166
Specific Gravity 0.74
Solubility Information Solubility in water: 5g/L water (20°C)
Viscosity 0.8 mPa.s (20°C)
Formula Weight 129.24
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Diisobutylamine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Diisobutylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H301-Toxic if swallowed.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'